Merck
  • Amino acid efflux in the isolated perfused rat pancreas: trans-stimulation by extracellular amino acids.
Amino acid efflux in the isolated perfused rat pancreas: trans-stimulation by extracellular amino acids.
The Journal of physiology (1989-09-01)
G E Mann, P S Norman, I C Smith
ZUSAMMENFASSUNG
1. Epithelial uptake and efflux of the non-metabolized system A analogue 2-methylaminoisobutyric acid (MeAIB) and L-serine were studied in the isolated perfused rat pancreas using a dual tracer loading and wash-out technique. Uptakes of 2-[14C]MeAIB and L-[3H]serine were measured relative to D-[3H or 14C]mannitol (extracellular tracer) during a 20 min cell loading period. Maximal uptake for MeAIB (34 +/- 2%, n = 6) occurred within 2-3 min and decreased to 14 +/- 2% after 20 min tracer loading. Uptake for L-serine reached a maximum (62 +/- 4%, n = 7) within 1 min and decreased to 19 +/- 2% after 20 min tracer loading. 2. When tracer wash-out was monitored during subsequent perfusion of the preloaded pancreas with an isotope-free solution, D-mannitol predominantly cleared from a fast exchanging compartment (0.54 +/- 0.05 ml g-1, n = 9) with a time constant (Tfast) of 0.68 +/- 0.04 min. Although MeAIB and L-serine exhibited similar fast phases of wash-out, a much larger efflux occurred from a slowly exchanging pool with respective time constants (Tslow) of 15.47 +/- 0.45 min (n = 6) and 5.98 +/- 0.46 min (n = 7). 3. A rapid vascular challenge of the pancreas with 100 mM-L-serine transiently accelerated cellular efflux of 2-[14C]MeAIB and L-[3H]serine without affecting wash-out of D-[14C]mannitol. Tracer efflux following cell loading with 2-[14C]MeAIB or L-[3H]serine was not stimulated by a challenge with 100 mM-MeAIB. 4. The time course of amino acid evoked 2-[14C]MeAIB and L-[3H]serine efflux paralleled the extracellular dilution profile of a vascular stimulus, suggesting that the acceleration of efflux was due to trans-stimulation. 5. Trans-stimulation of 2-[14C]MeAIB and L-[3H]serine efflux by a further twenty-two naturally occurring and three synthetic amino acids was then examined. L-Proline, N-methyl-DL-alanine, L-lysine and D-lysine selectively stimulated MeAIB efflux. Efflux of both tracer amino acids was accelerated by aminoisobutyric acid (AIB), L-serine, L-alanine, L-cysteine, L-threonine, glycine, branched-chain and large neutral amino acids, but affected negligibly by D-alanine, beta-alanine, MeAIB, L-arginine, L-aspartate, L-glutamate, taurine or D-mannitol. 6. In summary, we have characterized amino acid exchange mechanisms in the isolated pancreas and conclude that efflux of intracellular amino acids from pancreatic acinar cells may be mediated by at least two transporters with differing specificity for extracellular amino acids.(ABSTRACT TRUNCATED AT 400 WORDS)