Merck
  • Thiourea-Derived Chelating Ligands and Their Organometallic Compounds: Investigations into Their Anticancer Activity.
Thiourea-Derived Chelating Ligands and Their Organometallic Compounds: Investigations into Their Anticancer Activity.
Molecules (Basel, Switzerland) (2020-08-17)
Kelvin K H Tong, Muhammad Hanif, James H Lovett, Katja Hummitzsch, Hugh H Harris, Tilo Söhnel, Stephen M F Jamieson, Christian G Hartinger
ZUSAMMENFASSUNG
Thiones have been investigated as ligands in metal complexes with catalytic and biological activity. We report the synthesis, characterization, and biological evaluation of a series of MII/III complexes of the general formulae [MII(cym)(L)Cl]X (cym = η6-p-cymene) or [MIII(Cp*)(L)Cl]X (Cp* = η5-pentamethylcyclopentadienyl), where X = Cl- or PF6-, and L represents heterocyclic derivatives of thiourea. The thiones feature a benzyl-triazolyl pendant and they act as bidentate ligands via N,S-coordination to the metal centers. Several derivatives have been investigated by single-crystal X-ray diffraction analysis. NMR investigations showed a counterion-dependent shift of several protons due to the interaction with the counterions. These NMR investigations were complemented with X-ray diffraction analysis data and the effects of different counterions on the secondary coordination sphere were also investigated by DFT calculations. In biological studies, the Ir benzimidazole derivative was found to accumulate in the cytoplasm and it was the most cytotoxic derivative investigated.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1-Methylbenzimidazol, 99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.