Merck
  • Metabolic profiling of synthetic cannabinoid 5F-ADB and identification of metabolites in authentic human blood samples via human liver microsome incubation and ultra-high-performance liquid chromatography/high-resolution mass spectrometry.
Metabolic profiling of synthetic cannabinoid 5F-ADB and identification of metabolites in authentic human blood samples via human liver microsome incubation and ultra-high-performance liquid chromatography/high-resolution mass spectrometry.
Rapid communications in mass spectrometry : RCM (2020-07-28)
Brandon C Presley, Marisol S Castaneto, Barry K Logan, Susan A Jansen-Varnum
ZUSAMMENFASSUNG
Indazole carboxamide synthetic cannabinoids, a prevalent class of recreational drugs, are a major clinical, forensic and public health challenge. One such compound, 5F-ADB, has been implicated in fatalities worldwide. Understanding its metabolism and distribution facilitates the development of laboratory assays to substantiate its consumption. Synthetic cannabinoid metabolites have been extensively studied in urine; studies identifying metabolites in blood are limited and no data on the metabolic stability (half-life, clearance and extraction ratio) of 5F-ADB have been published prior to this report. The in vitro metabolism of 5F-ADB was elucidated via incubation with human liver microsomes for 2 h at 37°C. Samples were collected at multiple time points to determine its metabolic stability. Upon identification of metabolites, authentic forensic human blood samples underwent liquid-liquid extraction and were screened for metabolites. Extracts were analyzed via ultra-high-performance liquid chromatography/quadrupole time-of-flight mass spectrometry (UHPLC/QTOFMS) operated in positive electrospray ionization mode. Seven metabolites were identified including oxidative defluorination (M1); carboxypentyl (M2); monohydroxylation of the fluoropentyl chain (M3.1/M3.2) and indazole ring system (M4); ester hydrolysis (M5); and ester hydrolysis with oxidative defluorination (M6). The half-life (3.1 min), intrinsic clearance (256.2 mL min-1  kg-1 ), hepatic clearance (18.6 mL min-1 kg-1 ) and extraction ratio (0.93) were determined for the first time. In blood, M1 was present in each sample as the most abundant substance; two samples contained M5; one contained 5F-ADB, M1 and M5. 5F-ADB is rapidly metabolized in HLM. 5F-ADB, M1 and M5 are pharmacologically active at the cannabinoid receptors (CB1 /CB2 ) and M1 and M5 may contribute to a user's impairment profile. The results demonstrate that it is imperative that synthetic cannabinoid assays screen for pharmacologically active metabolites, especially for drugs with short half-lives. The authors propose that M1 and M5 are appropriate markers to include in laboratory blood tests screening for 5F-ADB.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,2-Dimethylbuttersäure, 96%