Merck
Osteocalcin does not influence acute or chronic inflammation in human vascular cells.
Journal of cellular physiology (2019-09-25)
Sophie A Millar, Ieva Zala, Susan I Anderson, Saoirse E O'Sullivan
ZUSAMMENFASSUNG
Some human observational studies have suggested an anti-inflammatory role of osteocalcin (OCN). An inflammatory protocol using interferon-γ and tumor necrosis factor-α (10 ng/ml) was employed to examine the acute (24 hr) and chronic (144 hr) effects of uncarboxylated OCN (ucOCN) in commercial, primary, subcultured human aortic endothelial cells (HAEC), and human smooth muscle cells (HASMCs). The inflammatory protocol increased phosphorylation of intracellular signaling proteins (CREB, JNK, p38, ERK, AKT, STAT3, STAT5) and increased secretion of adhesion markers (vascular cell adhesion molecule-1, intracellular adhesion molecule-1, monocyte chemoattractant protein-1) and proinflammatory cytokines (interleukin-6 [IL-6], IL-8). After acute inflammation, there were no additive or reductive effects of ucOCN in either cell type. Following chronic inflammation, ucOCN did not affect cell responses, nor did it appear to have any pro- or anti-inflammatory effects when administered acutely or chronically on its own in either cell type. Additionally, ucOCN did not affect lipopolysaccharide (LPS)-induced acute inflammation in HAECs or HASMCs. The findings of this study do not support a causal role for OCN within the models of vascular inflammation chosen. Further confirmatory studies are warranted.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
MILLIPLEX MAP Multi-Pathway Magnetic Bead 9-Plex – Multiplex-Zellsignalisierungsassay