Merck
  • Synthesis and validation of novel cholesterol-based fluorescent lipids designed to observe the cellular trafficking of cationic liposomes.
Synthesis and validation of novel cholesterol-based fluorescent lipids designed to observe the cellular trafficking of cationic liposomes.
Bioorganic & medicinal chemistry letters (2015-08-06)
Bieong-Kil Kim, Young-Bae Seu, Jong-Soo Choi, Jong-Won Park, Kyung-Oh Doh
ZUSAMMENFASSUNG
Cholesterol-based fluorescent lipids with ether linker were synthesized using NBD (Chol-E-NBD) or Rhodamine B (Chol-E-Rh), and the usefulnesses as fluorescent probes for tracing cholesterol-based liposomes were validated. The fluorescent intensities of liposomes containing these modified lipids were measured and observed under a microscope. Neither compound interfered with the expression of GFP plasmid, and live cell images were obtained without interferences. Changes in the fluorescent intensity of liposomes containing Chol-E-NBD were followed by flow cytometry for up to 24h. These fluorescent lipids could be useful probes for trafficking of cationic liposome-mediated gene delivery.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Avanti
18:1-06:0 NBD PE, Avanti Research - A Croda Brand 810155C