Merck
  • Biochar counteracts nitrification inhibitor DMPP-mediated negative effect on spinach (Spinacia oleracea L.) growth.
Biochar counteracts nitrification inhibitor DMPP-mediated negative effect on spinach (Spinacia oleracea L.) growth.
Ecotoxicology and environmental safety (2020-02-01)
Jamal Sheikhi, Hossein Mirsyed Hosseini, Hassan Etesami, Aziz Majidi
ZUSAMMENFASSUNG
The use of nitrification inhibitors (NIs) such as 3,4-dimethylpyrazole phosphate (DMPP) has been suggested to diminish agricultural soil nitrate (NO3-) loss and increase nitrogen (N) use efficiency (NUE). However, the yield of ammonium (NH4+)-sensitive plants such as spinach (Spinacia oleracea L.) may be adversely affected by the application of NIs at high N levels and, on the other hand, the efficiency of the NIs may also be affected by soil amendments such as biochar. These two issues are still not adequately addressed. The aim of this study was to evaluate the effect of different N levels including DMPP or not in a calcareous soil with and without amendment of wheat straw biochar on spinach yield, NUE, nitrate concentration of spinach leaf, activity of enzymes nitrate reductase (NR) and nitrite reductase (NiR), and soil ammonium (NH4+) and NO3- concentration under greenhouse conditions. This experiment was carried out with different N rates factor at seven levels (un-fertilized, N0; fertilized with 50 mg N kg-1 soil, N50; fertilized with 75 mg N kg-1 soil, N75; fertilized with 100 mg N kg-1 soil, N100; fertilized with N50 + DMPP; fertilized with N75 + DMPP; and fertilized with N100 + DMPP) and biochar (BC) factor at two levels (0, 0%BC; and 2% (w/w), 2%BC) with six replications over a 56-day cultivation period of spinach. Results showed that the application of DMPP had no significant effect on the yield of spinach plant at low and medium levels of N (50 and 75 mg N kg-1 soil), but decreased the yield of this plant at the higher level of N (100 mg N kg-1 soil). However, application of BC decreased the negative effect of DMPP on spinach yield as the yield in spinach plants fertilized with N75 + DMPP and N100 + DMPP significantly increased. Both application of DMPP and addition of BC to soil decreased leaf NO3- concentration by 29.2% and 16.3% compared to control, respectively. Biochar compared to control decreased NR activity by 46.3%. With increasing N rate, NR and NiR activities increased, but DMPP decreased the activities of both enzymes. Biochar reduced the efficiency of DMPP as soil NH4+ concentration was higher in the treatments containing DMPP without BC at 56 days after planting. Biochar and DMPP could increase the quality of spinach plant through decreasing the leaf NO3- concentration. In general, wheat straw biochar counteracted DMPP-mediated negative effect on growth of spinach plant at high level of N by decreasing the efficiency of this inhibitor. These results provide the useful information for managing the application rate of N fertilizers including DMPP in biochar-amended soil.