- Expression of orexin A and its receptor 1 in the vestibular glands of the cattle genital tract.
Expression of orexin A and its receptor 1 in the vestibular glands of the cattle genital tract.
Anatomical record (Hoboken, N.J. : 2007) (2008-12-18)
Luigi Michele Pavone, Simona Tafuri, Luigi Avallone, Norma Staiano, Alfredo Vittoria
PMID19089887
ZUSAMMENFASSUNG
The hypothalamic peptide orexin A (oxA) binds specifically the G-protein-coupled orexin receptor 1 (ox1R). It is involved in many physiological functions including the regulation of food intake, sleep-wake cycle, arterial blood pressure, heart rate, and sexual behavior. The localization of oxA in adrenal glands, stomach, bowel, pancreas, and testis has recently been assessed. Here, we provide the first evidence for the expression of oxA and ox1R in the vestibular glands of mammalian genital tract.