Merck
  • Paracetamol inhibits Ca2+ permeant ion channels and Ca2+ sensitization resulting in relaxation of precontracted airway smooth muscle.
Paracetamol inhibits Ca2+ permeant ion channels and Ca2+ sensitization resulting in relaxation of precontracted airway smooth muscle.
Journal of pharmacological sciences (2019-12-18)
Yuan-Yuan Chen, Meng-Fei Yu, Xiao-Xue Zhao, Jinhua Shen, Yong-Bo Peng, Ping Zhao, Lu Xue, Weiwei Chen, Li-Qun Ma, Gangjian Qin, Jiapei Dai, Qing-Hua Liu
ZUSAMMENFASSUNG
The purpose of this study was to screen a bronchodilator from old drugs and elucidate the underlying mechanism. Paracetamol (acetaminophen) is a widely used analgesic and antipyretic drug. It has been reported that it inhibits the generation of prostaglandin and histamine, which play roles in asthma. These findings led us to explore whether paracetamol could be a potential bronchodilator. Paracetamol inhibited high K+- and acetylcholine (ACH)-induced precontraction of mouse tracheal and bronchial smooth muscles. Moreover, the ACH-induced contraction was partially inhibited by nifedipine (selective blocker of LVDCCs), YM-58483 (selective inhibitor of store-operated Ca2+ entry (SOCE), canonical transient receptor potential 3 (TRPC3) and TRPC5 channels) and Y-27632 (selective blocker of ROCK, a linker of the Ca2+ sensitization pathway). In single airway smooth muscle cells, paracetamol blocked the currents sensitive to nifedipine and YM-58483, and inhibited intracellular Ca2+ increases. In addition, paracetamol inhibited ACH-induced phosphorylation of myosin phosphatase target subunit 1 (MYPT1, another linker of the Ca2+ sensitization pathway). Finally, in vivo paracetamol inhibited ACH-induced increases of mouse respirator system resistance. Collectively, we conclude that paracetamol inhibits ASM contraction through blocking LVDCCs, SOCE and/or TRPC3 and/or TRPC5 channels, and Ca2+ sensitization. These results suggest that paracetamol might be a new bronchodilator.