Merck
Incomplete T-cell receptor-beta peptides target the mitochondrion and induce apoptosis.
Blood (2008-10-22)
Nir Shani, Hila Rubin-Lifshitz, Yifat Peretz-Cohen, Ketty Shkolnik, Vera Shinder, Michal Cohen-Sfady, Yaron Shav-Tal, Mira Barda-Saad, Dov Zipori
ZUSAMMENFASSUNG
The default pathway of cell-surface T-cell receptor (TCR) complex formation, and the subsequent transport to the membrane, is thought to entail endoplasmic reticulum (ER) localization followed by proteasome degradation of the unassembled chains. We show herein an alternative pathway: short, incomplete peptide versions of TCRbeta naturally occur in the thymus. Such peptides, which have minimally lost the leader sequence or have been massively truncated, leaving only the very C terminus intact, are sorted preferentially to the mitochondrion. As a consequence of the mitochondrial localization, apoptotic cell death is induced. Structure function analysis showed that both the specific localization and induction of apoptosis depend on the transmembrane domain (TMD) and associated residues at the COOH-terminus of TCR. Truncated forms of TCR, such as the short peptides that we detected in the thymus, may be products of protein degradation within thymocytes. Alternatively, they may occur through the translation of truncated mRNAs resulting from unfruitful rearrangement or from germline transcription. It is proposed that mitochondria serve as a subcellular sequestration site for incomplete TCR molecules.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-ZAP-70-Antikörper, Klon 2F3.2, clone 2F3.2, Upstate®, from mouse