Merck
  • LRPAP1 is a frequent proliferation-inducing antigen of BCRs of mantle cell lymphomas and can be used for specific therapeutic targeting.
LRPAP1 is a frequent proliferation-inducing antigen of BCRs of mantle cell lymphomas and can be used for specific therapeutic targeting.
Leukemia (2018-06-30)
Lorenz Thurner, Sylvia Hartmann, Natalie Fadle, Maria Kemele, Theresa Bock, Moritz Bewarder, Evi Regitz, Frank Neumann, Anna Nimmesgern, Lutz von Müller, Christiane Pott, Yoo-Jin Kim, Rainer Maria Bohle, Mariusz Wasik, Stephen J Schuster, Martin-Leo Hansmann, Klaus-Dieter Preuss, Michael Pfreundschuh
ZUSAMMENFASSUNG
The predominant usage of VH4-34 and V3-21 and reports of stereotyped CDR3s suggest a shared antigenic target of B-cell receptors (BCR) from mantle cell lymphomas (MCL). To identify the target antigens of MCL-BCRs, BCRs from 21 patients and seven MCL cell lines were recombinantly expressed and used for antigen screening. The BCRs from 8/21 patients and 2/7 MCL cell lines reacted specifically with the autoantigen low-density lipoprotein receptor-related protein-associated protein 1 (LRPAP1). High-titered and light chain-restricted anti-LRPAP1 serum antibodies were found in MCL patients, but not in controls. LRPAP1 induced proliferation by BCR pathway activation, while an LRPAP1-ETA' toxin-conjugate specifically killed MCL cells with LRPAP1-specific BCRs. Our results suggest a role of LRPAP1 in lymphomagenesis and maintenance of a considerable proportion of MCL cases by chronic autoantigenic stimulation, likely evolving from a chronic autoreactive B-cell response. Importantly, LRPAP1 can be used for a novel therapeutic approach that targets MCL with LRPAP1-reactive BCRs with high specificity.