Merck
  • Antimicrobial properties of the novel bacterial isolate Paenibacilllus sp. SMB1 from a halo-alkaline lake in India.
Antimicrobial properties of the novel bacterial isolate Paenibacilllus sp. SMB1 from a halo-alkaline lake in India.
Scientific reports (2019-08-11)
Harjodh Singh, Manpreet Kaur, Manoj Jangra, Sunita Mishra, Hemraj Nandanwar, Anil Kumar Pinnaka
ZUSAMMENFASSUNG
Antibiotic-resistance is ever growing burden on our society for the past many years. Many synthetic chemistry approaches and rational drug-design have been unable to pace up and tackle this problem. Natural resources, more specifically, the microbial diversity, on the other hand, make a traditional and still the best platform to search for new chemical scaffolds and compounds. Here, we report the antimicrobial characteristics of novel bacterial isolate from a salt lake in India. We screened the bacterial isolates for their inhibitory activity against indicator bacteria and found that four novel species were able to prevent the growth of test strains studied in vitro. Further, we characterized one novel species (SMB1T = SL4-2) using polyphasic taxonomic approaches and also purified the active ingredient from this bacterium. We successfully characterized the antimicrobial compound using mass spectroscopy and amino acid analysis. We also allocated two novel biosynthetic gene clusters for putative bacteriocins and one novel non-ribosomal peptide gene cluster in its whole genome. We concluded that the strain SMB1T belonged to the genus Paenibacilllus with the pairwise sequence similarity of 98.67% with Paenibacillus tarimensis DSM 19409T and we proposed the name Paenibacillus sambharensis sp. nov. The type strain is SMB1T (=MTCC 12884 = KCTC 33895T).