Merck
  • A model of cholinergic suppression of hippocampal ripples through disruption of balanced excitation/inhibition.
A model of cholinergic suppression of hippocampal ripples through disruption of balanced excitation/inhibition.
Hippocampus (2018-11-13)
Eric D Melonakos, John A White, Fernando R Fernandez
ZUSAMMENFASSUNG
Sharp wave-ripples (140-220 Hz) are patterns of brain activity observed in the local field potential of the hippocampus which are present during memory consolidation. As rodents switch from memory consolidation to memory encoding behaviors, cholinergic inputs to the hippocampus from neurons in the medial septum-diagonal band of Broca cause a marked reduction in ripple incidence. The mechanism for this disruption in ripple power is not fully understood. In isolated neurons, the major effect of cholinergic input on hippocampal neurons is depolarization of the membrane potential, which affects both hippocampal pyramidal neurons and inhibitory interneurons. Using an existing model of ripple-frequency oscillations that includes both pyramidal neurons and interneurons, we investigated the mechanism whereby depolarizing inputs to these neurons can affect ripple power and frequency. We observed that ripple power and frequency are maintained, as long as inputs to pyramidal neurons and interneurons are balanced. Preferential drive to pyramidal neurons or interneurons, however, affects ripple power and can disrupt ripple oscillations by pushing ripple frequency higher or lower. Thus, an imbalance in drive to pyramidal neurons and interneurons provides a means whereby cholinergic input can suppress hippocampal ripples.