Merck
  • Light emission from the Fe2+ -EDTA-ascorbic acid-H2 O2 system strongly enhanced by plant phenolic acids.
Light emission from the Fe2+ -EDTA-ascorbic acid-H2 O2 system strongly enhanced by plant phenolic acids.
Luminescence : the journal of biological and chemical luminescence (2019-01-25)
Michal Nowak, Wiesław Tryniszewski, Agata Sarniak, Anna Włodarczyk, Piotr J Nowak, Dariusz Nowak
ZUSAMMENFASSUNG
Oxidative reactions can result in the formation of electronically excited species that undergo radiative decay depending on electronic transition from the excited state to the ground state with subsequent ultra-weak photon emission (UPE). We investigated the UPE from the Fe2+ -EDTA (ethylenediaminetetraacetic acid)-AA (ascorbic acid)-H2 O2 (hydrogen peroxide) system with a multitube luminometer (Peltier-cooled photon counter, spectral range 380-630 nm). The UPE, of 92.6 μmol/L Fe2+ , 185.2 μmol/L EDTA, 472 μmol/L AA, 2.6 mmol/L H2 O2 , reached 1217 ± 118 relative light units during 2 min measurement and was about two times higher (P < 0.001) than the UPE of incomplete systems (Fe2+ -AA-H2 O2 , Fe2+ -EDTA-H2 O2 , AA-H2 O2 ) and medium alone. Substitution of Fe2+ with Cr2+ , Co2+ , Mn2+ or Cu2+ as well as of EDTA with EGTA (ethylene glycol-bis(β-aminoethyl ether)-N,N,N',N'-tetraacetic acid) or citrate powerfully inhibited UPE. Experiments with scavengers of reactive oxygen species (dimethyl sulfoxide, mannitol, sodium azide, superoxide dismutase) revealed the dependence of UPE only on hydroxyl radicals. Dimethyl sulfoxide at the concentration of 0.74 mmol/L inhibited UPE by 79 ± 4%. Plant phenolics (ferulic, chlorogenic and caffec acids) at the concentration of 870 μmol/L strongly enhanced UPE by 5-, 13.9- and 46.8-times (P < 0.001), respectively. It is suggested that augmentation of UPE from Fe2+ -EDTA-AA-H2 O2 system can be applied for detection of these phytochemicals.