Merck
  • Direct measurement of the mechanical properties of a chromatin analog and the epigenetic effects of para-sulphonato-calix[4]arene.
Direct measurement of the mechanical properties of a chromatin analog and the epigenetic effects of para-sulphonato-calix[4]arene.
Scientific reports (2019-04-11)
Yannick Tauran, Momoko Kumemura, Mehmet C Tarhan, Grégoire Perret, Florent Perret, Laurent Jalabert, Dominique Collard, Hiroyuki Fujita, Anthony W Coleman
ZUSAMMENFASSUNG
By means of Silicon Nano Tweezers (SNTs) the effects on the mechanical properties of λ-phage DNA during interaction with calf thymus nucleosome to form an artificial chromatin analog were measured. At a concentration of 100 nM, a nucleosome solution induced a strong stiffening effect on DNA (1.1 N m-1). This can be compared to the effects of the histone proteins, H1, H2A, H3 where no changes in the mechanical properties of DNA were observed and the complex of the H3/H4 proteins where a smaller increase in the stiffness is observed (0.2 N m-1). Para-sulphonato-calix[4]arene, SC4, known for epigenetic activity by interacting specifically with the lysine groups of histone proteins, was studied for its effect on an artificial chromatin. Using a microfluidic SNT device, SC4 was titrated against the artificial chromatin, at a concentration of 1 mM in SC4 a considerable increase in stiffness, 15 N m-1, was observed. Simultaneously optical microscopy showed a physical change in the DNA structure between the tips of the SNT device. Electronic and Atomic Force microscopy confirmed this structural re-arrangement. Negative control experiments confirmed that these mechanical and physical effects were induced neither by the acidity of SC4 nor through nonspecific interactions of SC4 on DNA.