Merck
  • Acid anhydride coated carbon nanodots: activated platforms for engineering clicked (bio)nanoconstructs.
Acid anhydride coated carbon nanodots: activated platforms for engineering clicked (bio)nanoconstructs.
Nanoscale (2019-04-10)
Mariano Ortega-Muñoz, Paula Vargas-Navarro, Fernando Hernandez-Mateo, Alfonso Salinas-Castillo, Luis Fermin Capitan-Vallvey, Simona Plesselova, Rafael Salto-Gonzalez, M Dolores Giron-Gonzalez, Francisco Javier Lopez-Jaramillo, Francisco Santoyo-Gonzalez
ZUSAMMENFASSUNG
Activated carbon nanodots functionalized with acid anhydride groups (AA-CNDs) are prepared by one-pot water-free green thermolysis of citric acid. As a proof of concept of their capabilities as appealing and versatile platforms for accessing engineering nanoconstructs, the as-prepared AA-CNDs have been reacted to yield clickable CNDs. Their click bioconjugation with relevant recognizable complementary clickable sugars has led to multivalent CND-based glyconanoparticles that are non-toxic and biorecognizable. The accessibility and intrinsic reactivity of AA-CNDs expand the current toolbox of covalent surface grafting methodologies and provide a wide range of potential applications for engineering (bio)nanoconstructs.