Merck
  • Uptake and accumulation of emerging contaminants in soil and plant treated with wastewater under real-world environmental conditions in the Al Hayer area (Saudi Arabia).
Uptake and accumulation of emerging contaminants in soil and plant treated with wastewater under real-world environmental conditions in the Al Hayer area (Saudi Arabia).
The Science of the total environment (2018-10-28)
Yolanda Picó, Rodrigo Alvarez-Ruiz, Ahmed H Alfarhan, Mohamed A El-Sheikh, Samy M Alobaid, Damià Barceló
ZUSAMMENFASSUNG
In arid and semi-arid areas the use of treated wastewater for crop irrigation and other agricultural practices, such as the use of pesticides, increase the number of emerging contaminants (ECs) in crops. Hazards of these practices to human being are largely unknown since there are few studies yet covering a short range of compounds and most of them under non-realistic conditions. This study aims at assessing this problem that will become global soon in an area of Saudi Arabia heavily affected by the reuse of treated wastewater and pesticide in order to ascertain its scale. The novelty of the study relays in the large number of ECs covered and the variety of crops (cabbage, barley, green beans, eggplants, chili, tomato and zucchini) analysed. Extraction procedure developed provided an appropriate extraction yield (up to 50% of the compounds were recovered within a 70-120% range), with good repeatability (relative standard deviations below 20% in most cases) and sensitivity (LOQ < 25 ng g-1) for the model compounds. Determination by liquid chromatography quadrupole time-of-flight (LC-QqTOF-MS) is able to identify >2000 contaminants. Sixty-four ECs were identified in wastewater but of the sixty-four compounds, six pharmaceuticals (atenolol, caffeine, carbamazepine and its metabolites 10,11-epoxycarbamazepine, gemfibrozil, and naproxen) and seven pesticides (acetamiprid, atrazine deethyl, azoxystrobin, bupirimate, diazinon, malathion, pirimicarb and some of their metabolites) were detected in plants. Furhermore, one metabolite of the ibuprofen (not detected in water or soil), the ibuprofen hexoside was also found in plants. Up to our knowledge, this study demonstrate for the first time the accumulation of ECs in crops irrigated with treated wastewater under real non-controlled environmental conditions.