Merck
  • Leber hereditary optic neuropathy mutations in the ND6 subunit of mitochondrial complex I affect ubiquinone reduction kinetics in a bacterial model of the enzyme.
Leber hereditary optic neuropathy mutations in the ND6 subunit of mitochondrial complex I affect ubiquinone reduction kinetics in a bacterial model of the enzyme.
The Biochemical journal (2007-09-27)
Jukka Pätsi, Marko Kervinen, Moshe Finel, Ilmo E Hassinen
ZUSAMMENFASSUNG
LHON (Leber hereditary optic neuropathy) is a maternally inherited disease that leads to sudden loss of central vision at a young age. There are three common primary LHON mutations, occurring at positions 3460, 11778 and 14484 in the human mtDNA (mitochondrial DNA), leading to amino acid substitutions in mitochondrial complex I subunits ND1, ND4 and ND6 respectively. We have now examined the effects of ND6 mutations on the function of complex I using the homologous NuoJ subunit of Escherichia coli NDH-1 (NADH:quinone oxidoreductase) as a model system. The assembly level of the NDH-1 mutants was assessed using electron transfer from deamino-NADH to the 'shortcut' electron acceptor HAR (hexammine ruthenium), whereas ubiquinone reductase activity was determined using DB (decylubiquinone) as a substrate. Mutant growth in minimal medium with malate as the main carbon source was used for initial screening of the efficiency of energy conservation by NDH-1. The results indicated that NuoJ-M64V, the equivalent of the common LHON mutation in ND6, had a mild effect on E. coli NDH-1 activity, while nearby mutations, particularly NuoJ-Y59F, NuoJ-V65G and NuoJ-M72V, severely impaired the DB reduction rate and cell growth on malate. NuoJ-Met64 and NuoJ-Met72 position mutants lowered the affinity of NDH-1 for DB and explicit C-type inhibitors, whereas NuoJ-Y59C displayed substrate inhibition by oxidized DB. The results are compatible with the notion that the ND6 subunit delineates the binding cavity of ubiquinone substrate, but does not directly take part in the catalytic reaction. How these changes in the enzyme's catalytic properties contribute to LHON pathogenesis is discussed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Nicotinamide hypoxanthine dinucleotide sodium salt, ≥92%
Sigma-Aldrich
Nicotinamid-hypoxanthin-dinucleotid, reduzierte Form, Natriumsalz, Grade I, ≥95%