- Role of glycogen synthase kinase-3 in the phosphatidylinositol 3-Kinase/Akt cell survival pathway.
Role of glycogen synthase kinase-3 in the phosphatidylinositol 3-Kinase/Akt cell survival pathway.
The Journal of biological chemistry (1998-08-01)
M Pap, G M Cooper
PMID9685326
ZUSAMMENFASSUNG
Growth factor-dependent survival of a variety of mammalian cells is dependent on the activation of phosphatidylinositol (PI) 3-kinase and its downstream effector, the protein kinase Akt. Glycogen synthase kinase-3 (GSK-3) has been previously identified as a physiological target of Akt, which is inhibited by phosphorylation, so we have investigated the role of GSK-3 in cell survival. Overexpression of catalytically active GSK-3 induced apoptosis of both Rat-1 and PC12 cells, whereas dominant-negative GSK-3 prevented apoptosis following inhibition of PI 3-kinase. GSK-3 thus plays a critical role in regulation of apoptosis and represents a key downstream target of the PI 3-kinase/Akt survival signaling pathway.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phospho-Glycogen Synthase Peptide-2, Phospho-Glycogen Synthase Peptide-2 primarily used in Kinase Assays.