Merck
  • Sensitive Screening Method for Determination of Pyrethroids in Chicken Eggs and Various Meat Samples by Gas Chromatography and Electron Capture Detection.
Sensitive Screening Method for Determination of Pyrethroids in Chicken Eggs and Various Meat Samples by Gas Chromatography and Electron Capture Detection.
Journal of agricultural and food chemistry (2018-09-13)
Valeria Nardelli, Francesco Casamassima, Giuseppe Gesualdo, Donghao Li, Wadir M V Marchesiello, Donatella Nardiello, Maurizio Quinto
ZUSAMMENFASSUNG
A sensitive and reproducible screening analytical method is described for the determination of six pyrethroids (phenothrin, permethrin, cyfluthrin, cypermethrin, deltamethrin, fenvalerate) in egg and meat samples by gas chromatography and electron capture detection (GC-ECD). A fast cleanup procedure, based on solid-phase extraction has been used, ensuring reduced solvent consumption and time and allowing the simultaneous preparation of multiple sample extracts. Under the optimal chromatographic conditions, an efficient separation was obtained with a total analysis time of less than 60 min, including the extraction-purification steps. Good responses for the six pyrethroids were obtained in a range of 50-500 μg L-1, with linear coefficients higher than 0.9992. Instrumental limits of detection were between 0.22 and 0.63 μg L-1, corresponding to 0.04 and 0.13 μg kg-1 in the matrix. Detection limits in chicken eggs and various meat samples, calculated on spiked samples, were in the range 0.05-0.25 μg kg-1 and 0.07-0.23 μg kg-1, respectively. The validation results confirmed that the proposed GC-ECD method can be used as a reliable screening tool for the determination of pyrethroids in official check analyses. The method was extensively validated following the European directives, demonstrating its conformity in terms of selectivity, sensitivity, recovery, precision, and measurement uncertainty.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Phenothrin, PESTANAL®, analytical standard