Merck
Subcellular organization of UBE3A in neurons.
The Journal of comparative neurology (2016-06-25)
Alain C Burette, Matthew C Judson, Susan Burette, Kristen D Phend, Benjamin D Philpot, Richard J Weinberg
ZUSAMMENFASSUNG
Ubiquitination regulates a broad array of cellular processes, and defective ubiquitination is implicated in several neurological disorders. Loss of the E3 ubiquitin-protein ligase UBE3A causes Angelman syndrome. Despite its clinical importance, the normal role of UBE3A in neurons is still unclear. As a step toward deciphering its possible functions, we performed high-resolution light and electron microscopic immunocytochemistry. We report a broad distribution of UBE3A in neurons, highlighted by concentrations in axon terminals and euchromatin-rich nuclear domains. Our findings suggest that UBE3A may act locally to regulate individual synapses while also mediating global, neuronwide influences through the regulation of gene transcription. J. Comp. Neurol. 525:233-251, 2017. © 2016 Wiley Periodicals, Inc.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoclonal Anti-UBE3A antibody produced in mouse, clone 3E5, purified immunoglobulin, buffered aqueous solution