Skip to Content
Merck
All Photos(1)

Key Documents

SML1538

Sigma-Aldrich

NMDI14

≥97% (HPLC)

Synonym(s):

4,5-Dimethyl-2-[[2-(1,2,3,4-tetrahydro-6,7-dimethyl-3-oxo-2-quinoxalinyl)acetyl]amino]-3-thiophenecarboxylic acid ethyl ester, Ethyl 2-{[(6,7–dimethyl–3-oxo-1,2,3,4-tetrahydro-2-quinoxalinyl)acetyl]amino}-4,5-dimethyl-3-thiophenecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H25N3O4S
CAS Number:
Molecular Weight:
415.51
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 0.2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

[s]1c(c(c(c1C)C)C(=O)OCC)NC(=O)CC2Nc3c(cc(c(c3)C)C)NC2=O

InChI key

QRLPSNLBUMVXBL-UHFFFAOYSA-N

Application

NMDI14 has been used as a nonsense-mediated mRNA decay (NMD) inhibitor:
  • to study its effects on differentiating cardiomyocytes
  • to study its effects on zebrafish pdzk1 gene-knockout embryos
  • to analyze its effects on the apoptosis of colorectal cancer cells

Biochem/physiol Actions

NMDI14 is a potent nonsense-mediated RNA decay (NMD) inhibitor. NMDI14 targets a pocket in the SMG7 protein and disrupts SMG7–UPF1 interactions. NMDI14 restores of full-length p53 protein activity in in cells with premature termination codons (PTC) mutated p53.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Bordonaro et al.
Journal of Cancer, 10(2), 287-292 (2019-02-06)
Deregulated Wnt signaling initiates most cases of colorectal cancer (CRC). Butyrate, a product of dietary fiber, hyperactivates Wnt signaling, resulting in induction of CRC cell apoptosis, which may in part explain the protective action of fiber. Nonsense mediated decay (NMD)
Xinyun Chen et al.
Nature communications, 12(1), 89-89 (2021-01-06)
The RNA-binding protein QKI belongs to the hnRNP K-homology domain protein family, a well-known regulator of pre-mRNA alternative splicing and is associated with several neurodevelopmental disorders. Qki is found highly expressed in developing and adult hearts. By employing the human
Greg H P Ngo et al.
Nature communications, 12(1), 3849-3849 (2021-06-24)
DNA-RNA hybrid structures have been detected at the vicinity of DNA double-strand breaks (DSBs) occurring within transcriptional active regions of the genome. The induction of DNA-RNA hybrids strongly affects the repair of these DSBs, but the nature of these structures
Jiaheng Xie et al.
Investigative ophthalmology & visual science, 62(3), 29-29 (2021-03-23)
The human PDZK1 gene is located in a genomic susceptibility region for neurodevelopmental disorders. A genome-wide association study identified links between PDZK1 polymorphisms and altered visual contrast sensitivity, an endophenotype for schizophrenia and autism spectrum disorder. The PDZK1 protein is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service