Skip to Content
Merck
All Photos(1)

Key Documents

SML3433

Sigma-Aldrich

NVP-TNKS656

≥98% (HPLC)

Synonym(s):

; N-Cyclopropylmethyl-2-[4-(4-methoxybenzoyl)piperidin-1-yl]-N-[(4-oxo-4,5,7,8-tetrahydro-4H-pyrano[4,3-d]pyrimidin-2-yl)methyl]acetamide, N-​(Cyclopropylmethyl)​-​4-​(4-​methoxybenzoyl)​-​N-​[(3,​5,​7,​8-​tetrahydro-​4-​oxo-​4H-​pyrano[4,​3-​d]​pyrimidin-​2-​yl)​methyl]​-​piperidineacetamide, NVP-TNKS 656, TNKS656

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H34N4O5
CAS Number:
Molecular Weight:
494.58
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N4(CCC(CC4)C(=O)c5ccc(cc5)OC)CC(=O)N(CC3CC3)Cc1[nH][c](c2c(n1)CCOC2)=O

InChI

1S/C27H34N4O5/c1-35-21-6-4-19(5-7-21)26(33)20-8-11-30(12-9-20)16-25(32)31(14-18-2-3-18)15-24-28-23-10-13-36-17-22(23)27(34)29-24/h4-7,18,20H,2-3,8-17H2,1H3,(H,28,29,34)

InChI key

DYGBNAYFDZEYBA-UHFFFAOYSA-N

Biochem/physiol Actions

NVP-TNKS656 is an orally active, highly potent and selective tankyrase inhibitor. NVP-TNKS656 is a potent antagonist of Wnt pathway that reverts resistance to PI3K and AKT inhibitors in treatment of colorectal cancer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service