Skip to Content
Merck
All Photos(1)

Documents

EHU081751

Sigma-Aldrich

MISSION® esiRNA

targeting human DNM1L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTTTTCACCCAACGTTGTCAATTTGACACTTGTGGATTTGCCAGGAATGACCAAGGTGCCTGTAGGTGATCAACCTAAGGATATTGAGCTTCAAATCAGAGAGCTCATTCTTCGGTTCATCAGTAATCCTAATTCCATTATCCTCGCTGTCACTGCTGCTAATACAGATATGGCAACATCAGAGGCACTTAAAATTTCAAGAGAGGTAGATCCAGATGGTCGCAGAACCCTAGCTGTAATCACTAAACTTGATCTCATGGATGCGGGTACTGATGCCATGGATGTATTGATGGGAAGGGTTATTCCAGTCAAACTTGGAATAATTGGAGTAGTTAACAGGAGCCAGCTAGATATTAACAACAAGAAGAGTGTAACTGATTCAATCCGTGATGAGTATGCTTTTCTTCAAAAGAAATATCCATCTCTGGCCAATAGAAA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicole L Mancini et al.
Cellular and molecular gastroenterology and hepatology, 11(2), 551-571 (2020-09-30)
Adherent-invasive Escherichia coli are implicated in inflammatory bowel disease, and mitochondrial dysfunction has been observed in biopsy specimens from patients with inflammatory bowel disease. As a novel aspect of adherent-invasive E coli-epithelial interaction, we hypothesized that E coli (strain LF82)
Maria Manczak et al.
Human molecular genetics, 28(2), 177-199 (2018-09-22)
The purpose of our study was to better understand the effects of mitochondrial-division inhibitor 1 (Mdivi-1) on mitochondrial fission, mitochondrial biogenesis, electron transport activities and cellular protection. In recent years, researchers have found excessive mitochondrial fragmentation and reduced fusion in
Dongjoon Kim et al.
Cells, 9(7) (2020-07-16)
Diabetic retinopathy is a prevalent microvascular complication characterized by apoptotic vascular cell loss in the retina. Previous studies have shown that high glucose (HG)-induced mitochondrial fragmentation plays a critical role in promoting retinal vascular cell apoptosis. Here, we investigated whether
Unbin Chae et al.
Bioscience, biotechnology, and biochemistry, 83(3), 409-416 (2018-11-27)
Microglial activation is known to be an important event during innate immunity, but microglial inflammation is also thought to play a role in the etiology of neurodegenerative diseases. Recently, it was reported that autophagy could influence inflammation and activation of
S Xu et al.
Cell death & disease, 4, e540-e540 (2013-03-16)
Mitochondria are critical targets in the hepatotoxicity of cadmium (Cd). Abnormal mitochondrial dynamics have been increasingly implicated in mitochondrial dysfunction in pathophysiological conditions. Therefore, our study aimed to investigate the effects and underlying mechanism of Cd on mitochondrial dynamics during

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service