Key Documents
CLS6625
Corning® Step-R Repeating Pipettor Syringe Tips
volume (12.5 mL), non-sterile
Synonym(s):
pipet tips
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
manufacturer/tradename:
Corning 6625
volume:
(12.5 mL)
sterility:
non-sterile
packaging:
pack of 100 ea
sterility
non-sterile
packaging
pack of 100 ea
manufacturer/tradename
Corning 6625
volume
(12.5 mL)
General description
  • Ideal for viscous, dense and volatile liquides
  • Minimum increments printed on the syringe itself
  • Accurately deliver selected dose
  • Made from high quality polypropylene and polyethylene
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)