Key Documents
Z376892
Nichiryo digital hand-held dispenser
dispensing volume 2.0-10.0 mL
About This Item
UNSPSC Code:
41121500
NACRES:
NB.51
Pricing and availability is not currently available.
feature
electronic: no
single-channel
packaging
pack of 1 ea
manufacturer/tradename
Nichiryo 00-21DG-10
dispensing volume measuring range
2.0-10.0 mL
volume
2 × 10 mL
General description
This light weight dispenser allows for continuous dispensing of solutions. Glass cylinder and plunger are protected from breakage by plastic housings. All parts which come in direct contact with the solution are autoclavable. Provides accuracy of ±1% and precision of <0.2%.
Choose from one of the most recent versions:
Certificates of Analysis (COA)