Skip to Content
Merck
All Photos(1)

Key Documents

N18

88112301, Mouse x Rat Hybridoma hybrid, Neuronal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
Mouse x Rat Hybridoma hybrid
growth mode:
Adherent
karyotype:
Not specified
morphology:
Neuronal
products:
Not specified
receptors:
Not specified
relevant disease(s):
cancer

Product Name

N18, 88112301

biological source

Mouse x Rat Hybridoma hybrid

growth mode

Adherent

karyotype

Not specified

morphology

Neuronal

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse neuroblastoma x Rat glioma hybrid

Cell Line Description

Neuroblastoma x glioma hybrid was formed by Sendai virus-induced fusion of the mouse neuroblastoma clone N18.

Application

Hybridisation, fusion partner and virus studies: Sendai virus

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Selection of the right fusion partner cell line is critical to obtain desired attributes of the hybridoma. Explore hybridoma fusion partners cell lines.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service