H5
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
biological source
rat liver
packaging
tube of 5 μg 94101905-DNA-5UG
pkg of vial of cells 94101905-1VL
growth mode
Adherent
karyotype
Modal no. 52 (50-55)
morphology
Fusiform
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Rat hepatoma
Cell Line Description
H5 is a subclone of H4-II-E-C3 (Sigma Catalogue number. 85061112) which originated from the Reuber H-35 hepatoma induced by chemical carcinogenesis in an A x C rat. The dedifferentiated H5 cells express high levels of glucose-6-phosphate dehydrogenase and exhibit a population doubling time of 12 hours. Differentiated variants are H4IIEC3 (85061112), Fao and others.
Application
Toxicity studies; negative control for differentiated hepatoma
Culture Medium
Coon′s modified Ham′s F12 (or Kaign′s modified Ham′s F12 + 45mg/L ascorbic acid + 18mg/L Inositol) + 2mM Glutamine + 5% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:50 to 1:100 i.e. seeding at 1-2x1,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service