Key Documents
Z742669
Artis Tweezer
Style MM-SA
About This Item
UNSPSC Code:
27112105
NACRES:
MA.01
Pricing and availability is not currently available.
feature
tip style (fine points.)
manufacturer/tradename
Aven 18023ARS
technique(s)
PCR: suitable
L
125 mm (4.75 in.)
General description
Aven′s innovative Artis Tweezers have set a new precedent for precision. Our cutting edge design features a angled bend at the end of the tip, which provides a softer overall grip with a large surface area that reduces hand fatigue, enhancing productivity.
The durable tweezers are protected by an attractive ESD safe coating, and the highly polished stainless steel tips with rounded edges allow operators to handle very fine wire, miniature parts, electrical components, and other delicate materials in a confined space without risk of damage.
Application
Excellent for general assembly, with strong fine points.
Choose from one of the most recent versions:
Certificates of Analysis (COA)