Merck
Prognostic roles of mRNA expression of notch receptors in non-small cell lung cancer.
Oncotarget (2017-01-07)
Jianwen Xiong, Xiaoqiang Zhang, Xianglai Chen, Yiping Wei, De-Guo Lu, Yun-Wei Han, Jianjun Xu, Dongliang Yu
RÉSUMÉ
Notch signalling is aberrantly activated in human non-small cell lung cancer (NSCLC). Nevertheless, the prognostic roles of mRNA expression of four Notch receptors in NSCLC patients remain elusive. In this report, we reported the prognostic roles of Notch receptors in a total of 1,926 NSCLC patients through "The Kaplan-Meier plotter" (KM plotter) database which is capable to assess the effect of 22,277 genes on survival of NSCLC patients. We found that mRNA high expression level of Notch1 was associated with better overall survival (OS) for all NSCLC patients, hazard ratio (HR) 0.78 (0.69-0.89), p=0.00019, better OS in adenocarcinoma (Ade) patients, HR 0.59 (0.46-0.75), p=1.5e-05, as well as in squamous cell carcinoma (SCC) patients, HR 0.78 (0.62-0.99), p=0.044. mRNA high expression levels of Notch2 and Notch3 were associated with worsen OS for all NSCLC patients, as well as in Ade, but not in SCC patients. mRNA high expression level of Notch4 was not found to be associated with to OS for all NSCLC patients. In addition, mRNA high expression levels of Notch2, Notch3, but Notch4 are significantly associated with the NSCLC patients who have different smoking status. Our results indicate that mRNA expression of Notch receptors may have distinct prognostic values in NSCLC patients. These results will benefit for developing tools to accurately predict the prognosis of NSCLC patients.