Merck
A large conformational change of the translocation ATPase SecA.
Proceedings of the National Academy of Sciences of the United States of America (2004-07-17)
Andrew R Osborne, William M Clemons, Tom A Rapoport
RÉSUMÉ
The ATPase SecA mediates the posttranslational translocation of a wide range of polypeptide substrates through the SecY channel in the cytoplasmic membrane of bacteria. We have determined the crystal structure of a monomeric form of Bacillus subtilis SecA at a 2.2-A resolution. A comparison with the previously determined structures of SecA reveals a nucleotide-independent, large conformational change that opens a deep groove similar to that in other proteins that interact with diverse polypeptides. We propose that the open form of SecA represents an activated state.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
n-Octyl β-D-maltoside, ≥99.0% (HPLC)