Merck
Fixation of atmospheric CO
Dalton transactions (Cambridge, England : 2003) (2017-07-27)
Utsab Manna, Santanu Kayal, Soham Samanta, Gopal Das
RÉSUMÉ
A meta-phenylenediamine-based disubstituted bis-urea receptor L1 with electron-withdrawing 3-chloro and electron-donating 4-methylphenyl terminals has been established as a potential system to fix and efficiently capture atmospheric CO
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
3-Chloro-4-methylphenyl isocyanate, 98%