Merck
Caspase 8 promotes peripheral localization and activation of Rab5.
The Journal of biological chemistry (2008-11-01)
Vicente A Torres, Ainhoa Mielgo, Daniela Barilà, Deborah H Anderson, Dwayne Stupack
RÉSUMÉ
Caspase 8 is a cysteine protease that initiates apoptotic signaling via the extrinsic pathway in a manner dependent upon association with early endosomes. Previously, we identified caspase 8 as an effector of migration, promoting motility in a manner dependent upon phosphorylation on Tyr-380 by Src family kinases and its subsequent association with Src homology 2 domain-containing proteins. Here we demonstrate the regulation of the small GTPase Rab5, which mediates early endosome formation, homotypic fusion, and maturation by caspase 8. Regulation requires the Tyr-380 phosphorylation site but not caspase proteolytic activity. Tyr-380 is essential for interaction with the Src homology 2 domains of p85alpha, a multifunctional adaptor for phosphatidylinositol 3-kinase, that possesses Rab-GAP activity. Interaction between caspase 8 and p85alpha promotes Rab5 GTP loading, alters endosomal trafficking, and results in the accumulation of Rab5-positive endosomes at the edge of the cell. Conversely, caspase 8-dependent GTP loading of Rab5 is overcome by increased expression of p85alpha in a Rab-GAP-dependent manner. Thus, we demonstrate a novel function for caspase 8 as a modulator of p85alpha Rab-GAP activity and endosomal trafficking.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anticorps monoclonal anti-β-actine antibody produced in mouse, clone AC-74, ascites fluid