Merck
Nitroxide free radical clearance in the live rat monitored by radio-frequency CW-EPR and PEDRI.
Physics in medicine and biology (1998-08-14)
M Alecci, I Seimenis, S J McCallum, D J Lurie, M A Foster
RÉSUMÉ
The use of RF (100 to 300 MHz) PEDRI and CW-EPR techniques allows the in vivo study of large animals such as whole rats and rabbits. Recently a PEDRI instrument was modified to also allow CW-EPR spectroscopy with samples of similar size and under the same experimental conditions. In the present study, this CW-EPR and PEDRI apparatus was used to assess the feasibility of the detection of a pyrrolidine nitroxide free radical (2,2,5,5,-tetramethylpyrrolidine-1-oxyl-3-carboxylic acid, PCA) in the abdomen of rats. In particular, we have shown that after the PCA administration (4 mmol kg(-1) b.w.): (i) the PCA EPR linewidth does not show line broadening due to concentration effects; (ii) a similar PCA up-take phase is observed by EPR and PEDRI; and (iii) the PCA half-lives in the whole abdomen of rats measured with the CW-EPR (T1/2=26+/-4 min, mean+/-sd, n=10) and PEDRI (T1/2=29+/-4 min, mean+/-sd, n=4) techniques were not significantly different (p > 0.05). These results show, for the first time, that information about PCA pharmacokinetics obtained by CW-EPR is the same as that from PEDRI under the same experimental conditions.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
3-Carboxy-PROXYL, 98%