Merck
Synthesis and secretory expression of hybrid antimicrobial peptide CecA-mag and its mutants in Pichia pastoris.
Experimental biology and medicine (Maywood, N.J.) (2012-03-02)
Xiuqing Wang, Mingxing Zhu, Aijun Zhang, Fengqin Yang, Puyan Chen
RÉSUMÉ
The hybrid peptide CA(1-7)-M(2-12) gene was designed according to the N-terminal 1-7 amino acid sequence of the antimicrobial peptide cecropin A (CA) and the N-terminal 2-12 amino acid sequence of maganin (M) and synthesized using Pichia pastoris preferred codons. The gene was cloned into pPICZαA and transformed into the P. pastoris recipient bacterium SMD1168, regulated by the alcohol oxidase (AOX). Expression of the cecA-mag hybrid antimicrobial peptide (MW, 1.9 kDa) revealed broad-spectrum antibiotic activity and to the ability to inhibit growth of most G(-) and G(+) bacteria. Three mutants of cecA-mag were designed and synthesized by recombination polymerase chain reaction site-directed mutagenesis to investigate the relationship between the structure and function of this antimicrobial peptide. The inhibition titers of these mutants against Staphylococcus aureus were evaluated using the agar diffusion method. Under the conditions of the same concentration and volume, the bacteriostatic diameters of three cecA-mag mutants were 1.2, 1.2 and 1.5 times, respectively, compared with the diameters of wild-type cecA-mag.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Cecropin A, ≥97% (HPLC), powder