Merck
  • Development and validation of HPLC method for simultaneous estimation of piperine and guggulsterones in compound Unani formulation (tablets) and a nanoreservoir system.
Development and validation of HPLC method for simultaneous estimation of piperine and guggulsterones in compound Unani formulation (tablets) and a nanoreservoir system.
Biomedical chromatography : BMC (2012-01-04)
Y T Kamal, S Mohammed Musthaba, Mhaveer Singh, Rabea Parveen, Sayeed Ahmad, Sanjula Baboota, Imran Ali, Khalid M Siddiqui, S M Arif Zaidi
RÉSUMÉ
An attempt has been made to develop and validate a simultaneous HPLC method for novel approach of drug release via oil-in-water (o/w) nanoemulsion formulation and Habb-e-Khardal Unani tablet containing piperine and guggul sterones E and Z as main ingredients. Nanoemulsion was prepared by titration method using sefsol-218 as an oily phase, cremophor-EL as a surfactant, transcutol as a co-surfactant and distilled water as an aqueous phase. The formulation was optimized on the basis of thermodynamic stability and dispersibilty test. The nanoformulation was evaluated for particle size, surface morphology, electrical conductivity and viscosity determination. The in vitro dissolution was carried out by dialysis bag method. Drugs were quantified using an HPLC method developed in-house with a C(18) column as stationary phase and acetonitrile and water as mobile phase at λ(max) of 240 nm. The optimized formulation showed higher drug release, lower droplet size and less viscosity as compared with the conventional Habb-e-Khardal Unani tablet. The present study illustrated the potential of nanoemulsion dosage form in improving biopharmaceutic performance of piperine and guggul sterone. The HPLC method was also found to be quite sufficient for the routine quality control of formulations containing piperine and guggul sterone E and Z as ingredients and also for in vitro drug release studies.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
(Z)-Guggulsterone, ≥89% (HPLC), powder
Sigma-Aldrich
(E)-Guggulsterone, ≥95% (HPLC), powder