Merck
Analysis of SUMO1-conjugation at synapses.
eLife (2017-06-10)
James A Daniel, Benjamin H Cooper, Jorma J Palvimo, Fu-Ping Zhang, Nils Brose, Marilyn Tirard
RÉSUMÉ
SUMO1-conjugation of proteins at neuronal synapses is considered to be a major post-translational regulatory process in nerve cell and synapse function, but the published evidence for SUMO1-conjugation at synapses is contradictory. We employed multiple genetic mouse models for stringently controlled biochemical and immunostaining analyses of synaptic SUMO1-conjugation. By using a knock-in reporter mouse line expressing tagged SUMO1, we could not detect SUMO1-conjugation of seven previously proposed synaptic SUMO1-targets in the brain. Further, immunostaining of cultured neurons from wild-type and SUMO1 knock-out mice showed that anti-SUMO1 immunolabelling at synapses is non-specific. Our findings indicate that SUMO1-conjugation of synaptic proteins does not occur or is extremely rare and hence not detectable using current methodology. Based on our data, we discuss a set of experimental strategies and minimal consensus criteria for the validation of SUMOylation that can be applied to any SUMOylation substrate and SUMO isoform.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-mGluR7 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-GluR6/7 Antibody, clone NL9, rabbit monoclonal, culture supernatant, clone NL904, Upstate®