- Unimolecular micelles based on hydrophobically derivatized hyperbranched polyglycerols: ligand binding properties.
Unimolecular micelles based on hydrophobically derivatized hyperbranched polyglycerols: ligand binding properties.
Biomacromolecules (2008-02-06)
Rajesh Kumar Kainthan, Clement Mugabe, Helen M Burt, Donald E Brooks
PMID18247528
RÉSUMÉ
This paper discusses the binding and release properties of hydrophobically modified hyperbranched polyglycerol-polyethylene glycol copolymers that were originally developed as human serum albumin (HSA) substitutes. Their unimolecular micellar nature in aqueous solution has been proven by size measurements and other spectroscopic methods. These polymers aggregate weakly in solution, but the aggregates are broken down by low shear forces or by encapsulating a hydrophobic ligand within the polymer. The small molecule binding properties of these polymers are compared with those of HSA. The preliminary in vitro paclitaxel release studies showed very promising sustained drug release characteristics achieved by these unimolecular micelles.