Merck
Autophagic degradation of lamins facilitates the nuclear egress of herpes simplex virus type 1.
The Journal of cell biology (2018-12-28)
Aykut Turan, Linda Grosche, Adalbert Krawczyk, Petra Mühl-Zürbes, Christina Drassner, Alexandra Düthorn, Mirko Kummer, Mike Hasenberg, Sylvia Voortmann, Holger Jastrow, Jan Dörrie, Niels Schaft, Max Kraner, Katinka Döhner, Beate Sodeik, Alexander Steinkasserer, Christiane Silke Heilingloh
RÉSUMÉ
Dendritic cells (DCs) are crucial for the induction of potent antiviral immune responses. In contrast to immature DCs (iDCs), mature DCs (mDCs) are not permissive for infection with herpes simplex virus type 1 (HSV-1). Here, we demonstrate that HSV-1 infection of iDCs and mDCs induces autophagy, which promotes the degradation of lamin A/C, B1, and B2 in iDCs only. This in turn facilitates the nuclear egress of progeny viral capsids and thus the formation of new infectious particles. In contrast, lamin protein levels remain stable in HSV-1-infected mDCs due to an inefficient autophagic flux. Elevated protein levels of KIF1B and KIF2A in mDCs inhibited lamin degradation, likely by hampering autophagosome-lysosome fusion. Therefore, in mDCs, fewer progeny capsids were released from the nuclei into the cytosol, and fewer infectious virions were assembled. We hypothesize that inhibition of autophagic lamin degradation in mDCs represents a very powerful cellular counterstrike to inhibit the production of progeny virus and thus viral spread.