Merck
Derivation of Mouse Haploid Trophoblast Stem Cells.
Cell reports (2019-01-10)
Tongtong Cui, Liyuan Jiang, Tianda Li, Fei Teng, Guihai Feng, Xuepeng Wang, Zhengquan He, Lu Guo, Kai Xu, Yihuan Mao, Leyun Wang, Xuewei Yuan, Liu Wang, Wei Li, Qi Zhou
RÉSUMÉ
Trophoblast stem (TS) cells are increasingly used as a model system for studying placentation and placental disorders. However, practical limitations of genetic manipulation have posed challenges for genetic analysis using TS cells. Here, we report the generation of mouse parthenogenetic haploid TS cells (haTSCs) and show that supplementation with FGF4 and inhibition of Rho-associated protein kinase (ROCK) enable the maintenance of their haploidy and developmental potential. The resulting haTSCs have 20 chromosomes, exhibit typical expression features of TS cells, possess the multipotency to differentiate into specialized trophoblast cell types, and can chimerize E13.5 and term placentas. We also demonstrate the capability of the haTSCs to undergo genetic manipulation and facilitate genome-wide screening in the trophoblast lineage. We expect that haTSCs will offer a powerful tool for studying functional genomics and placental biology.