Key Documents
HAM59000-25
Hamilton® Microlab® 600 instrument syringe
model, 1725 BFP, volume 250 μL
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.01
Matériaux
Gastight Borosilicate glass/PTFE syringe
Caractéristiques
model 1725 BFP
Fabricant/nom de marque
Hamilton 59000-25
Paramètres
10-115 °C temp. range (50-239 °F)
500 psig pressure
D.I. × D.E.
2.30 mm × 7.75 mm
Volume
250 μL
Informations légales
Hamilton is a registered trademark of Hamilton Co.
Microlab is a registered trademark of Hamilton Co.