Z740230
TLC Aluminum Sheets
L × W 20 cm × 20 cm, silica gel 60 matrix, binder, Organic Polymer, fluorescent indicator: no
Synonym(s):
TLC Silica gel 60 W, TLC Silica gel 60 W, 25 Aluminium sheets 20 x 20 cm
About This Item
Recommended Products
material
aluminum support
silica gel 60 matrix
Quality Level
feature
binder Organic Polymer
fluorescent indicator: no
packaging
pkg of 25 ea
manufacturer/tradename
EMD Millipore® 1.16487.0001
technique(s)
thin layer chromatography (TLC): suitable
L × W
20 cm × 20 cm
layer thickness
140-160 μm
particle size
10-12 μm
pore size
60 Å medium pore diameter
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Application of TLC-Spectrodensitometric and Chemometric Methods for Determination of Momenta Cream: A Comparative Study Applied on Ternary Mixture.: The research compares TLC-spectrodensitometric and chemometric methods for simultaneous determination of components in Momenta cream using aluminum TLC plates. The study validates these methods as effective for analyzing pharmaceutical formulations (Salem et al., 2021).
Packaging
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service