HS120265
Parafilm® M Sealing Film
film W × L 508 mm × 16 m, natural
About This Item
Recommended Products
material
natural
polyolefin (Thermoplastic)
sterility
non-sterile
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS120265
Parafilm PM998
film W × L
508 mm × 16 m
General description
Features and Benefits
- Protect the integrity of research applications with a tight moisture-barrier that holds a strong seal and doesn′t release from glassware, despite off-gassing
- Reduce the risk of tampered data with a covering that is odorless, colorless and semi-transparent
- Prevents moisture and volume loss due to material composition and tight seal
- Flexible and self-sealing material allows it to cling around irregular shapes and surfaces
- Stretches more than 200% of original length
- Offers good resistance against acids, alkalis and some organic solvents
- Resistant up to 48 hours against many polar substances such as saline solutions, inorganic acids and alkaline solutions
- Disposable
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)