Skip to Content
Merck
All Photos(1)

Documents

AXYTXLF10RS

MAXYMum Recovery® pipette filter tips, for Pipetman® P2/P10 and Eppendorf® Research/Reference pipettors, ultramicro, extra long

size 0.1-10 μL, pkg of 5x960tips/cs, racked, clear, sterile

Synonym(s):

MAXYMum Recovery® pipette filter tips, for Pipetman® P2/P10 and Eppendorf® Research/Reference pipettors, ultramicro, extra long

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.02

sterility

sterile

manufacturer/tradename

Axygen Inc TXLF-10-R-S
Corning

packaging

pkg of 5x960tips/cs

size

0.1-10 μL

color

clear

Looking for similar products? Visit Product Comparison Guide

Other Notes

Browse all pipette tips

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Maxymum Recovery is a registered trademark of Corning, Inc.
Pipetman is a registered trademark of Gilson S.A.S. LLC

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service