
MAXYMum Recovery® pipette filter tips, for Pipetman® P2/P10 and Eppendorf® Research/Reference pipettors, ultramicro, extra long

Synonym(s):
MAXYMum Recovery® pipette filter tips, for Pipetman® P2/P10 and Eppendorf® Research/Reference pipettors, ultramicro, extra long
Products
Product Category
Manufacturer
Color
Sterility
Showing 1-3 of 3
Compare
Product Number
Description
Pricing