E4658
Diethylene glycol diethyl ether
reagent grade, ≥98%
Synonym(s):
2-Ethoxyethyl ether, Bis(2-ethoxyethyl) ether, Diethyldiglycol
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
(C2H5OCH2CH2)2O
CAS Number:
Molecular Weight:
162.23
Beilstein:
1699259
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
grade:
reagent grade
Assay:
≥98%
bp:
180-190 °C (lit.)
vapor pressure:
0.5 mmHg ( 20 °C)
Recommended Products
grade
reagent grade
Quality Level
vapor density
5.6 (vs air)
vapor pressure
0.5 mmHg ( 20 °C)
Assay
≥98%
form
liquid
expl. lim.
6.2 %
refractive index
n20/D 1.412 (lit.)
bp
180-190 °C (lit.)
mp
-68 °C
density
0.909 g/mL at 25 °C (lit.)
SMILES string
CCOCCOCCOCC
InChI
1S/C8H18O3/c1-3-9-5-7-11-8-6-10-4-2/h3-8H2,1-2H3
InChI key
RRQYJINTUHWNHW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
152.6 °F - closed cup
Flash Point(C)
67 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service