Skip to Content
Merck
All Photos(2)

Key Documents

90834

Supelco

Bradykinin acetate salt

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H73N15O11 · xC2H4O2
CAS Number:
Molecular Weight:
1060.21 (free base basis)
EC Number:
MDL number:
UNSPSC Code:
85151701
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

analyte chemical class(es)

peptides

technique(s)

HPLC: suitable

impurities

≤10 wt. % water

color

white

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

N2([C@@H](CCC2)C(=O)NCC(=O)N[C@@H](Cc5ccccc5)C(=O)N[C@@H](CO)C(=O)N3[C@@H](CCC3)C(=O)N[C@@H](Cc4ccccc4)C(=O)N[C@@H](CCCN=C(N)N)C(=O)O)C(=O)[C@H]1N(CCC1)C(=O)[C@@H](N)CCCN=C(N)N

InChI

1S/C50H73N15O11/c51-32(16-7-21-56-49(52)53)45(72)65-25-11-20-39(65)47(74)64-24-9-18-37(64)43(70)58-28-40(67)59-34(26-30-12-3-1-4-13-30)41(68)62-36(29-66)46(73)63-23-10-19-38(63)44(71)61-35(27-31-14-5-2-6-15-31)42(69)60-33(48(75)76)17-8-22-57-50(54)55/h1-6,12-15,32-39,66H,7-11,16-29,51H2,(H,58,70)(H,59,67)(H,60,69)(H,61,71)(H,62,68)(H,75,76)(H4,52,53,56)(H4,54,55,57)/t32-,33-,34-,35-,36-,37-,38-,39-/m0/s1

InChI key

QXZGBUJJYSLZLT-FDISYFBBSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg

Application

Bradykinin acetate salt may be used as a precursor for the preparation of bradykinin (BK) prior to its determination from acidic aqueous solutions by electromembrane extraction (EME) followed by analysis using high performance liquid chromatography (HPLC) with ultraviolet (UV) detector.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Activates pain receptors; induces smooth muscle contraction; activates Ca2+-dependent nitric oxide synthetase; increases capillary permeability.
Bradykinin is a pro-inflammatory peptide that acts through G-protein-coupled receptors. It activates sphingolipid metabolism. Fibroblasts treated with bradykinin produce a rapid and important increase in ceramide which is followed by a transient rise in sphingosine content. The peptide also increases ceramide cellular content by rapidly mobilizing neutral glycolipids. Bradykinin plays an important role in the regulation of fluid and electrolyte balance, smooth muscle contraction, vasodilation and capillary permeability. The bradykinin potentiators, which possess no bradykinin activity themselves, enhance the activity of bradykinin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electromembrane extraction of peptides-Fundamental studies on the supported liquid membrane.
Seip KF, et al.
Journal of Separation Science, 34(23), 3410-3417 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service