Skip to Content
Merck
All Photos(2)

Key Documents

860374C

Avanti

16:0-d31-18:1 PE

1-palmitoyl-d31-2-oleoyl-sn-glycero-3-phosphoethanolamine, chloroform

Synonym(s):

110921

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H45NO8PD31
CAS Number:
Molecular Weight:
749.19
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (860374C-10mg)
pkg of 1 × 2.5 mL (860374C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860374C

concentration

10 mg/mL (860374C-10mg)
10 mg/mL (860374C-25mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([

InChI

1S/C39H76NO8P/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-39(42)48-37(36-47-49(43,44)46-34-33-40)35-45-38(41)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2/h17-18,37H,3-16,19-36,40H2,1-2H3,(H,43,44)/b18-17-/t37-/m1/s1/i2D3,4D2,6D2,8D2,10D2,12D2,14D2,16D2,19D2,21D2,23D2,25D2,27D2,29D2,31D2

InChI key

FHQVHHIBKUMWTI-UAXXPTAZSA-N

General description

Deuterated fatty acids experience exchange of the deuteriums on the alpha carbon to the carbonyl, i.e., C2 position, and will therefore be a mixture of compounds that are fully deuterated and partially deuterated at that position.

Application

16:0-d31-18:1 PE has been used as an internal lipid standard for the lipid extraction of mice tracheal epithelial cells and human bronchial samples.

Packaging

5 mL Clear Glass Sealed Ampule (860374C-10mg)
5 mL Clear Glass Sealed Ampule (860374C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipid analysis of airway epithelial cells for studying respiratory diseases
Zehethofer N, et al.
Chromatographia, 78(5-6), 403-413 (2015)
Andres T Cavazos et al.
Chemistry and physics of lipids, 231, 104910-104910 (2020-06-04)
Among the structurally diverse collection of lipids that comprise the membrane lipidome, polyunsaturated phospholipids are particularly vulnerable to oxidation. The role of α-tocopherol (vitamin E) is to protect this influential class of membrane phospholipid from oxidative damage. Whether lipid-lipid interactions
Adrian Paz Ramos et al.
Biochimica et biophysica acta. Biomembranes, 1862(5), 183201-183201 (2020-01-24)
Alkanes are known to promote the fluid lamellar (Lα)-to-inverted hexagonal (HII) phase transition of different phospholipids. In this work, we studied the interaction of decane and tetradecane with self-assemblies formed of 1-palmitoyl-2-oleoyl-sn-glycero-phosphoethanolamine (POPE), using sequential 2H and 31P solid-state NMR spectroscopy.
Kotaro Hama et al.
Journal of lipid research, 61(4), 523-536 (2020-02-23)
X-linked adrenoleukodystrophy (X-ALD) is an inherited disorder caused by deleterious mutations in the ABCD1 gene. The ABCD1 protein transports very long-chain FAs (VLCFAs) from the cytosol into the peroxisome where the VLCFAs are degraded through β-oxidation. ABCD1 dysfunction leads to
Nicole Harmouche et al.
Biophysical journal, 115(6), 1033-1044 (2018-09-10)
A synergistic enhancement of activities has been described for the amphipathic cationic antimicrobial peptides magainin 2 and PGLa when tested in antimicrobial assays or in biophysical experiments using model membranes. In the presence of magainin 2, PGLa changes from an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service