Skip to Content
Merck
All Photos(3)

Key Documents

840045X

Avanti

Brain PI(4)P

Avanti Research - A Croda Brand 840045X

Synonym(s):

L-α-phosphatidylinositol-4-phosphate (Brain, Porcine) (ammonium Salt):methanol:water (20:9:1)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (840045X-1mg)
pkg of 1 × 1 mL (840045X-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840045X

concentration

1 mg/mL (840045X-1mg)
5 mg/mL (840045X-5mg)

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP(O[C@H]1[C@H](O)[C@@H](O)[C@H](OP(O)([O-])=O)[C@@H](O)[C@H]1O)([O-])=O)(OC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.[NH4+].[NH4+]

InChI

1S/C47H84O16P2.2H3N/c1-3-5-7-9-11-13-15-17-19-20-22-24-26-28-30-32-34-36-41(49)61-39(37-59-40(48)35-33-31-29-27-25-23-21-18-16-14-12-10-8-6-4-2)38-60-65(57,58)63-47-44(52)42(50)46(43(51)45(47)53)62-64(54,55)56;;/h11,13,17,19,22,24,28,30,39,42-47,50-53H,3-10,12,14-16,18,20-21,23,25-27,29,31-38H2,1-2H3,(H,57,58)(H2,54,55,56);2*1H3/b13-11-,19-17-,24-22-,30-28-;;/t39-,42+,43?,44?,45?,46?,47-;;/m1../s1

InChI key

HJHASIMOAXACIC-CVKYTLMISA-N

General description

Phosphatidylinositol 4-phosphate (PI4P) is a precursor of phosphatidylinositol (4,5)-bisphosphate [PI(4,5)P2]. It is located in the membranes of Golgi apparatus.

Application

Brain PI(4)P has been used for molecular docking. It has also been used for the preparation of liposome for co-floatation.

Packaging

5 mL Clear Glass Sealed Ampule (840045X-1MG)
5 mL Clear Glass Sealed Ampule (840045X-5MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Eyes,Central nervous system, Liver,Kidney

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

RIM C2B domains target presynaptic active zone functions to PIP2-containing membranes
de Jong A, et al.
Neuron, 98(2), 335-349 (2018)
The RNA-binding site of poliovirus 3C protein doubles as a phosphoinositide-binding domain
Shengjuler D, et al.
Structure, 25(12), 1875-1886 (2017)
PI4P and PI (4, 5) P2 are essential but independent lipid determinants of membrane identity
Hammond GRV, et al.
Science, 337(6095), 727-730 (2012)
Dieter R Klopfenstein et al.
Molecular biology of the cell, 15(8), 3729-3739 (2004-05-25)
UNC-104 (KIF1A) is a kinesin motor that transports synaptic vesicles from the neuronal cell body to the terminal. Previous in vitro studies have shown that a Dictyostelium relative of UNC-104 transports liposomes containing acidic phospholipids, but whether this interaction is
Jitendra Kumar et al.
PLoS genetics, 6(11), e1001200-e1001200 (2010-11-17)
UNC-104/KIF1A is a Kinesin-3 motor that transports synaptic vesicles from the cell body towards the synapse by binding to PI(4,5)P(2) through its PH domain. The fate of the motor upon reaching the synapse is not known. We found that wild-type

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service