Z115460
Scienceware® Covamat® PE liner
roll W × L 30 in. × 50 ft
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122300
NACRES:
NB.35
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H24679-0000
roll W × L
30 in. × 50 ft
General description
Ribbed matting for lining lab tables, shelves, and trays; 0.01 in. thick with 0.03 in. thick ribs.
Legal Information
Covamat is a registered trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service