Merck
  • Cis/trans conformational equilibrium across the N-methylphenylalanine- N-methylphenylalanine peptide bond of arginine vasopressin analogs.
Cis/trans conformational equilibrium across the N-methylphenylalanine- N-methylphenylalanine peptide bond of arginine vasopressin analogs.
The journal of peptide research : official journal of the American Peptide Society (2004-04-23)
E Trzepałka, W Kowalczyk, B Lammek
RÉSUMÉ
Two cyclic analogs of vasopressin, -Pro-Arg-Gly-NH(2) (1) and -Pro-Arg-Gly-NH(2) (2) were synthesized by the solid phase method. Their structure was determined in aqueous solution by two-dimensional NMR techniques and simulated annealing algorithm from an extended template in X-PLOR. The total chemical shift correlation spectroscopy and rotating-frame Overhauser enhancement spectroscopy of the peptides displayed four distinct sets of residual proton resonances. This suggests that both analogs adopt four families of conformations in H(2)O/D(2)O (9 : 1) (one major and three minor species). In further analysis only signals of major species (M) and of one minor species (m(1)) were considered. The major species of both peptides include a trans peptide bond between the first and second residue, and a cis form between the second and third residue. In the minor species, all peptide bonds were found to exist in trans geometry.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
L-Phenylalanine methyl ester hydrochloride, 98%
Sigma-Aldrich
D-Phenylalanine methyl ester hydrochloride, 98%
Sigma-Aldrich
L-Phenylalanine methyl ester hydrochloride